Learn More
Yttrium(III) oxalate nonahydrate, 99.9% (REO), Thermo Scientific Chemicals
$196.01 - $196.01
Chemical Identifiers
CAS | 13266-82-5 |
---|---|
Molecular Formula | C6H18O21Y2 |
Molecular Weight (g/mol) | 604.001 |
MDL Number | MFCD00150502 |
InChI Key | CTXJKXXQOORSKU-UHFFFAOYSA-H |
PubChem CID | 131872179 |
IUPAC Name | oxalate;yttrium(3+);nonahydrate |
SMILES | C(=O)(C(=O)[O-])[O-].C(=O)(C(=O)[O-])[O-].C(=O)(C(=O)[O-])[O-].O.O.O.O.O.O.O.O.O.[Y+3].[Y+3] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA1456218
|
Thermo Scientific Chemicals
01456218 |
50 g |
Each for $196.01
|
|
|||||
Description
Yttrium oxalate is applied in ceramics, glass, and electronics. It is an important material for tri-bands rare Earth phosphors and yttrium-iron-garnets, which are very effective microwave filters.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13266-82-5 | |
604.001 | |
CTXJKXXQOORSKU-UHFFFAOYSA-H | |
oxalate;yttrium(3+);nonahydrate |
C6H18O21Y2 | |
MFCD00150502 | |
131872179 | |
C(=O)(C(=O)[O-])[O-].C(=O)(C(=O)[O-])[O-].C(=O)(C(=O)[O-])[O-].O.O.O.O.O.O.O.O.O.[Y+3].[Y+3] |
Specifications
(decomposition) | |
White | |
50 g | |
Y2(C2O4)3·9H2O | |
UN3288 | |
CTXJKXXQOORSKU-UHFFFAOYSA-H | |
oxalate;yttrium(3+);nonahydrate | |
131872179 | |
99.9% | |
(REO) |
13266-82-5 | |
Crystalline | |
C6H18O21Y2 | |
MFCD00150502 | |
Insoluble in water,moderately soluble in strong mineral acids | |
C(=O)(C(=O)[O-])[O-].C(=O)(C(=O)[O-])[O-].C(=O)(C(=O)[O-])[O-].O.O.O.O.O.O.O.O.O.[Y+3].[Y+3] | |
604.001 | |
604.01 (441.87 Anhydrous) | |
Yttrium(III) oxalate nonahydrate |
Safety and Handling
GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
H302+H312
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only