missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Yttrium(III) nitrate hydrate, REacton™, 99.99% (REO)

Manufacturer:  Alfa Aesar™ 1118759

 View more versions of this product

Catalog No. AA1118759

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Yttrium(III) nitrate hydrate
(REO)
MFCD00011469
14,10107
Soluble in water.
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.[Y+3]
292.933
274.92 (Anhydrous)
99.99%
Hygroscopic
140191-40-8
H2N3O10Y
UN1477
3no3.y.h2o, nitric acid, yttrium 3++ salt, nitric acid, yttrium iii salt, yttrium 3+ hydrate trinitrate, yttrium iii nitrate hydrate, yttrium iii nitrate hydrate, reacton, yttrium nitrate hydrate
WUVRZBFIXJWTGS-UHFFFAOYSA-N
yttrium(3+);trinitrate;hydrate
23169389
Crystalline Aggregates
Reacton™
5 x 100g

Chemical Identifiers

140191-40-8
292.933
WUVRZBFIXJWTGS-UHFFFAOYSA-N
23169389
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.[Y+3]
H2N3O10Y
MFCD00011469
3no3.y.h2o, nitric acid, yttrium 3++ salt, nitric acid, yttrium iii salt, yttrium 3+ hydrate trinitrate, yttrium iii nitrate hydrate, yttrium iii nitrate hydrate, reacton, yttrium nitrate hydrate
yttrium(3+);trinitrate;hydrate
Safety and Handling

Safety and Handling

GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H272-H315-H319-H335

DOTInformation : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.

EINECSNumber : 233-802-6

RTECSNumber : ZG3675000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents