missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Yttrium(III) nitrate hydrate, REacton™, 99.99% (REO)

Manufacturer:  Alfa Aesar™ 1118714

 View more versions of this product

Catalog No. AA1118714

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Yttrium(III) nitrate hydrate
(REO)
MFCD00011469
14,10107
WUVRZBFIXJWTGS-UHFFFAOYSA-N
yttrium(3+);trinitrate;hydrate
23169389
Crystalline Aggregates
Reacton™
25g
140191-40-8
H2N3O10Y
UN1477
3no3.y.h2o
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.[Y+3]
292.933
274.92 (Anhydrous)
99.99%
Hygroscopic

Chemical Identifiers

140191-40-8
292.933
WUVRZBFIXJWTGS-UHFFFAOYSA-N
23169389
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.[Y+3]
H2N3O10Y
MFCD00011469
3no3.y.h2o
yttrium(3+);trinitrate;hydrate
Safety and Handling

Safety and Handling

GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P221-P210-P220-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Keep/Store away from clothing/combustible materials.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.

TSCA : TSCA

Documents