missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Yttrium(III) nitrate hexahydrate, 99.9% (REO), Alfa Aesar™

Manufacturer:  Alfa Aesar 1289806

 View more versions of this product

Catalog No. AA1289806



Specifications

Specifications

Yttrium(III) nitrate hexahydrate
99.9% (REO)
MFCD00149940
QBAZWXKSCUESGU-UHFFFAOYSA-N
yttrium(3+);trinitrate;hexahydrate
166833
Crystalline
13494-98-9
H12N3O15Y
yttrium nitrate hexahydrate, yttrium iii nitrate hexahydrate, yttriumnitrat german, yttrium iii nitrate hexahydrate 1:3:6, yttrium trinitrate hexahydrate, nitric acid, yttrium 3+ salt, hexahydrate, nitric acid,yttrium 3+ salt, hexahydrate 8ci,9ci, yttriumnitrat, yttrium iii nitrate hexahydrate reo, ytterium nitrate
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Y+3]
383.008
383.01 (274.92 Anhydrous)
5g

Chemical Identifiers

13494-98-9
383.008
QBAZWXKSCUESGU-UHFFFAOYSA-N
166833
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Y+3]
H12N3O15Y
MFCD00149940
yttrium nitrate hexahydrate, yttrium iii nitrate hexahydrate, yttriumnitrat german, yttrium iii nitrate hexahydrate 1:3:6, yttrium trinitrate hexahydrate, nitric acid, yttrium 3+ salt, hexahydrate, nitric acid,yttrium 3+ salt, hexahydrate 8ci,9ci, yttriumnitrat, yttrium iii nitrate hexahydrate reo, ytterium nitrate
yttrium(3+);trinitrate;hexahydrate
Safety and Handling

Safety and Handling

EINECSNumber : 233-802-6

Documents