missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Yttrium(III) isopropoxide, 90+%

Manufacturer:  Alfa Aesar™ 1447706

 View more versions of this product

Catalog No. AA1447706



Specifications

Specifications

Yttrium(III) isopropoxide
200°C to 210°C (1mmHg)(sublimation)
5g
C9H24O3Y
UN2925
Soluble in hexane, THF, toluene
CC(C)O.CC(C)O.CC(C)O.[Y]
269.19
266.17
≥90%
Moisture Sensitive
160°C
2172-12-5
MFCD00015641
yttrium isopropoxide
NREVZTYRXVBFAQ-UHFFFAOYSA-N
propan-2-ol;yttrium
85308566
Powder

Chemical Identifiers

2172-12-5
269.19
NREVZTYRXVBFAQ-UHFFFAOYSA-N
85308566
CC(C)O.CC(C)O.CC(C)O.[Y]
C9H24O3Y
MFCD00015641
yttrium isopropoxide
propan-2-ol;yttrium
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H228
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable solid.

GHS P Statement
P210-P280-P303+P361+P353-P305+P351+P338-P310
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, CORROSIVE, ORGANIC, N.O.S.

Documents