missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Yttrium(III) isopropoxide, 20-25% w/w in toluene

Manufacturer:  Alfa Aesar™ 14524ND

 View more versions of this product

Catalog No. AA14524ND



Specifications

Specifications

Yttrium(III) isopropoxide
Aromatic
Yellow to Brown
2172-12-5
Y[OCH(CH3)2]3
UN3274
20 to 25% w/w in toluene
CC(C)O.CC(C)O.CC(C)O.[Y]
269.19
266.17
0.955g/mL
Air & Moisture Sensitive
≈25g
C9H24O3Y
MFCD00015641
yttrium isopropoxide
NREVZTYRXVBFAQ-UHFFFAOYSA-N
propan-2-ol;yttrium
85308566
Liquid

Chemical Identifiers

2172-12-5
269.19
NREVZTYRXVBFAQ-UHFFFAOYSA-N
85308566
CC(C)O.CC(C)O.CC(C)O.[Y]
C9H24O3Y
MFCD00015641
yttrium isopropoxide
propan-2-ol;yttrium
Safety and Handling

Safety and Handling

GHS H Statement
H225-H314-H318-H361-H373-H336
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
May cause drowsiness or dizziness.

GHS P Statement
P210-P280-P303+P361+P353-P305+P351+P338-P310
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ALCOHOLATES SOLUTION, N.O.S.

Documents