missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Yttrium(III) hexafluoro-2,4-pentanedionate

Manufacturer:  Alfa Aesar™ 1471706

 View more versions of this product

Catalog No. AA1471706



Specifications

Specifications

Yttrium(III) hexafluoro-2,4-pentanedionate
C15H6F18O6Y
Yttrium hexafluoroacetylacetonate
C(=C(C(F)(F)F)O)C(=O)C(F)(F)F.C(=C(C(F)(F)F)O)C(=O)C(F)(F)F.C(=C(C(F)(F)F)O)C(=O)C(F)(F)F.[Y]
713.084
710.1
166°C to 170°C
18911-76-7
MFCD00064764
QYKMGZLZIBNPQL-JVUUZWNBSA-N
(Z)-1,1,1,5,5,5-hexafluoro-4-hydroxypent-3-en-2-one;yttrium
50930692
Crystalline
5g

Chemical Identifiers

18911-76-7
713.084
QYKMGZLZIBNPQL-JVUUZWNBSA-N
50930692
C(=C(C(F)(F)F)O)C(=O)C(F)(F)F.C(=C(C(F)(F)F)O)C(=O)C(F)(F)F.C(=C(C(F)(F)F)O)C(=O)C(F)(F)F.[Y]
C15H6F18O6Y
MFCD00064764
Yttrium hexafluoroacetylacetonate
(Z)-1,1,1,5,5,5-hexafluoro-4-hydroxypent-3-en-2-one;yttrium
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents