Learn More
Alfa Aesar™ Yttrium(III) fluoride, anhydrous, REacton™, 99.9% (REO)
Manufacturer: Alfa Aesar™ 1365036
Specifications
Yttrium(III) fluoride | |
13709-49-4 | |
MFCD00049714 | |
145.9 | |
99.9% | |
Hygroscopic | |
500g |
Anhydrous | |
(REO) | |
UN3288 | |
Lump/Powder | |
Reacton™ | |
1387°C |
Chemical Identifiers
13709-49-4 | |
MFCD00049714 |
Safety and Handling
GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
TSCA : TSCA