missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Yttrium(III) chloride hydrate, REacton™, 99.99% (REO)

Manufacturer:  Alfa Aesar™ 1118530

 View more versions of this product

Catalog No. AA1118530



Specifications

Specifications

Yttrium(III) chloride hydrate
(REO)
YCl3·xH2O
14,10107
BWOAAAYNOWWMHL-UHFFFAOYSA-K
yttrium(3+);trichloride;hydrate
57347377
Crystalline Aggregates
Reacton™
Odorless
Pink to White
12741-05-8
Cl3H2OY
MFCD00149941
yttrium iii chloride hydrate, yttrium chloride, hydrate, acmc-1bq0g, ksc171o8f, yttrium chloride-water 1/3/1, yttrium 3+ ion hydrate trichloride
O.[Cl-].[Cl-].[Cl-].[Y+3]
213.271
195.26 (Anhydrous)
99.99%
2.18g/mL
Hygroscopic
250g

Chemical Identifiers

12741-05-8
213.271
BWOAAAYNOWWMHL-UHFFFAOYSA-K
57347377
O.[Cl-].[Cl-].[Cl-].[Y+3]
Cl3H2OY
MFCD00149941
yttrium iii chloride hydrate, yttrium chloride, hydrate, acmc-1bq0g, ksc171o8f, yttrium chloride-water 1/3/1, yttrium 3+ ion hydrate trichloride
yttrium(3+);trichloride;hydrate
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents