missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Yttrium(III) chloride hydrate, REacton™, 99.9% (REO)

$59.77 - $526.97

Chemical Identifiers

CAS 12741-05-8
Molecular Formula Cl3H2OY
Molecular Weight (g/mol) 213.271
MDL Number MFCD00149941
InChI Key BWOAAAYNOWWMHL-UHFFFAOYSA-K
Synonym acmc-1bq0g, ksc171o8f, yttrium 3+ ion hydrate trichloride, yttrium chloride, hydrate, yttrium chloride-water 1/3/1, yttrium iii chloride hydrate
PubChem CID 57347377
IUPAC Name yttrium(3+);trichloride;hydrate
SMILES O.[Cl-].[Cl-].[Cl-].[Y+3]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1118414
View Documents
Alfa Aesar™
1118414
25g Each for $59.77
 
AA1118422
View Documents
Alfa Aesar™
1118422
100g Each for $135.11
 
AA1118436
View Documents
Alfa Aesar™
1118436
500g Each for $526.97
 
Specifications

Chemical Identifiers

12741-05-8
213.271
BWOAAAYNOWWMHL-UHFFFAOYSA-K
57347377
O.[Cl-].[Cl-].[Cl-].[Y+3]
Cl3H2OY
MFCD00149941
acmc-1bq0g, ksc171o8f, yttrium 3+ ion hydrate trichloride, yttrium chloride, hydrate, yttrium chloride-water 1/3/1, yttrium iii chloride hydrate
yttrium(3+);trichloride;hydrate

Specifications

12741-05-8
MFCD00149941
acmc-1bq0g, ksc171o8f, yttrium 3+ ion hydrate trichloride, yttrium chloride, hydrate, yttrium chloride-water 1/3/1, yttrium iii chloride hydrate
BWOAAAYNOWWMHL-UHFFFAOYSA-K
yttrium(3+);trichloride;hydrate
57347377
Wet Crystalline Aggregates
Reacton™
Hygroscopic
Yttrium(III) chloride hydrate
Cl3H2OY
14,10107
Soluble in water.
O.[Cl-].[Cl-].[Cl-].[Y+3]
213.271
195.26 (Anhydrous)
99.9%
Odorless
25g
(REO)
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 233-801-0

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen