missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Yttrium(III) chloride hydrate, REacton™, 99.9% (REO)

Manufacturer:  Alfa Aesar™ 1118436

 View more versions of this product

Catalog No. AA1118436



Specifications

Specifications

Yttrium(III) chloride hydrate
(REO)
MFCD00149941
acmc-1bq0g, ksc171o8f, yttrium 3+ ion hydrate trichloride, yttrium chloride, hydrate, yttrium chloride-water 1/3/1, yttrium iii chloride hydrate
BWOAAAYNOWWMHL-UHFFFAOYSA-K
yttrium(3+);trichloride;hydrate
57347377
Wet Crystalline Aggregates
Reacton™
Hygroscopic
12741-05-8
Cl3H2OY
14,10107
Soluble in water.
O.[Cl-].[Cl-].[Cl-].[Y+3]
213.271
195.26 (Anhydrous)
99.9%
Odorless
500g

Chemical Identifiers

12741-05-8
213.271
BWOAAAYNOWWMHL-UHFFFAOYSA-K
57347377
O.[Cl-].[Cl-].[Cl-].[Y+3]
Cl3H2OY
MFCD00149941
acmc-1bq0g, ksc171o8f, yttrium 3+ ion hydrate trichloride, yttrium chloride, hydrate, yttrium chloride-water 1/3/1, yttrium iii chloride hydrate
yttrium(3+);trichloride;hydrate
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 233-801-0

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

Documents