Please login to your online account to display your discounted pricing

Yttrium(III) chloride hexahydrate, 99.999%, (trace metal basis), ACROS Organics™

Chemical Identifiers

Chemical Name or Material Yttrium(III) chloride hexahydrate
Name Note 99.999%
CAS 10025-94-2
Purity Grade For trace metal basis analysis
Assay ≥99.999% (metals basis)
Assay Percent Range 100.00%
Molecular Formula Cl3Y·6H2O
Linear Formula YCl3·6H2O
Formula Weight 303.35
MDL Number MFCD00149941
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC194470100

  SDS
thermo fisher scientific
194470100
Glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥99.999% (metals basis)
Physical Form Adhering Crystalline Powder
Formula Weight 303.35
Quantity 10g
Color White
Merck Index 14, 10107
TSCA TSCA
EINECS Number 233-801-0
RTECS Number ZG3150000
Purity Grade For trace metal basis analysis
Health Hazard 3 GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Harmful if inhaled.
Causes skin irritation.
Harmful in contact with skin.
Name Note 99.999%
Molecular Formula Cl3Y·6H2O
MDL Number MFCD00149941
CAS 10025-94-2
Health Hazard 1 GHS Signal Word: Warning
Packaging Glass bottle
Assay Percent Range 100.00%
Chemical Name or Material Yttrium(III) chloride hexahydrate
Linear Formula YCl3·6H2O

for ultra trace analysis

Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Harmful if inhaled.
Causes skin irritation.
Harmful in contact with skin.