missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Yttrium(III) acetate tetrahydrate, 99.9% (REO), Thermo Scientific Chemicals
In analytical chemistry
$116.08 - $393.63
Chemical Identifiers
CAS | 85949-60-6 |
---|---|
Molecular Formula | C6H17O10Y |
Molecular Weight (g/mol) | 338.10 |
MDL Number | MFCD00013048 |
InChI Key | AIQRTHPXPDTMBQ-UHFFFAOYSA-K |
IUPAC Name | yttrium(3+) triacetate tetrahydrate |
SMILES | O.O.O.O.[Y+3].CC([O-])=O.CC([O-])=O.CC([O-])=O |
Description
In analytical chemistry.Yttrium(III) acetate tetrahydrate is used in analytical chemistry and in catalysis. It is used in the preparation of yttrium trifluoride nanoparticles, which shows antibacterial properties.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
85949-60-6 | |
338.10 | |
AIQRTHPXPDTMBQ-UHFFFAOYSA-K | |
O.O.O.O.[Y+3].CC([O-])=O.CC([O-])=O.CC([O-])=O |
C6H17O10Y | |
MFCD00013048 | |
yttrium(3+) triacetate tetrahydrate |
Specifications
(decomposition) | |
85949-60-6 | |
50 g | |
Y(OOCCH3)3·4H2O | |
Soluble in hot water. | |
O.O.O.O.[Y+3].CC([O-])=O.CC([O-])=O.CC([O-])=O | |
338.10 | |
99.9% | |
Yttrium(III) acetate tetrahydrate |
White | |
Powder | |
C6H17O10Y | |
MFCD00013048 | |
AIQRTHPXPDTMBQ-UHFFFAOYSA-K | |
yttrium(3+) triacetate tetrahydrate | |
338.10 (266.04 Anhydrous) | |
1.5 g/mL | |
(REO) |
Safety and Handling
EINECSNumber : 245-612-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only