Please login to your online account to display your discounted pricing

Alfa Aesar™ Yttrium(III) 2-methoxyethoxide, 5% w/v in 2-methoxyethanol

Chemical Identifiers

Chemical Name or Material Yttrium(III) 2-methoxyethoxide
CAS 115668-57-0
Linear Formula Y(OCH2CH2OCH3)3
Formula Weight 314.165
MDL Number MFCD00156527
UN Number UN1188
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA41793AC

  SDS
alfa aesar™
41793AC
25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA41793AE

  SDS
alfa aesar™
41793AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 314.165
Quantity 25mL
Chemical Name or Material Yttrium(III) 2-methoxyethoxide
Physical Form Liquid
Linear Formula Y(OCH2CH2OCH3)3
MDL Number MFCD00156527
CAS 115668-57-0
UN Number UN1188
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H360-H226-H302-H312-H332
May damage fertility or the unborn child.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P201-P308+P313
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHYLENE GLYCOL MONOMETHYL ETHER
Concentration 5% w/v in 2-methoxyethanol
Sensitivity Moisture Sensitive
Safety and Handling

Danger

GHS P Statement
P201-P308+P313
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.

GHS H Statement
H360-H226-H302-H312-H332
May damage fertility or the unborn child.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.