Please login to your online account to display your discounted pricing

Alfa Aesar™ Yttrium(III) 2-ethylhexanoate, 99.8% (metals basis)

$131.99 - $131.99

Chemical Identifiers

Chemical Name or Material Yttrium(III) 2-ethylhexanoate
CAS 103470-68-4
Formula Weight 518.16
MDL Number MFCD00070441
Synonym 2-Ethylhexanoic acid yttrium(III) salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3666106

  SDS
alfa aesar™
3666106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3666114

  SDS
alfa aesar™
3666114
25g Each for $131.99

AA3666122

  SDS
alfa aesar™
3666122
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 518.16
Quantity 5g
Chemical Name or Material Yttrium(III) 2-ethylhexanoate
Assay 99.8%
Physical Form Powder
Synonym 2-Ethylhexanoic acid yttrium(III) salt
Health Hazard 2 GHS H Statement
H361
Suspected of damaging fertility or the unborn child.
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P280h
Wear protective gloves/clothing.
CAS 103470-68-4
Assay Percent Notes (metals basis)
MDL Number MFCD00070441
Safety and Handling

Warning

GHS P Statement
P280h
Wear protective gloves/clothing.

GHS H Statement
H361
Suspected of damaging fertility or the unborn child.