missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Yttrium Foil, 0.25mm (0.01 in.) thick, 0.999, Alfa Aesar™

Form: Approx. 0.7g/25 x 25mm

Manufacturer:  Alfa Aesar 13960FF

 View more versions of this product

Catalog No. AA13960FF



Specifications

Specifications

Yttrium
Rare Earth Oxides Content
Gray foil
Y
yttrium, elemental, yttrium-89, ion, ytrio, hydride, unii-58784xqc3y, yttrium, metal and compounds, ingot, foil, element
[Y]
Odorless
23993
Foil, 0.25mm (0.01 in.) thick
25 x 25mm
7440-65-5
99.90%
MFCD00011468
VWQVUPCCIRVNHF-UHFFFAOYSA-N
yttrium
88.906
CHEBI:33331
Gray

Chemical Identifiers

7440-65-5
88.906
VWQVUPCCIRVNHF-UHFFFAOYSA-N
23993
yttrium
Y
MFCD00011468
yttrium, elemental, yttrium-89, ion, ytrio, hydride, unii-58784xqc3y, yttrium, metal and compounds, ingot, foil, element
CHEBI:33331
[Y]
Documents