missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Yttrium Foil, 0.25mm (0.01 in.) thick, 0.999, Alfa Aesar™
Form: Approx. 0.7g/25 x 25mm
Manufacturer: Alfa Aesar 13960FI
Specifications
Yttrium | |
Rare Earth Oxides Content | |
Gray foil | |
99.90% | |
MFCD00011468 | |
VWQVUPCCIRVNHF-UHFFFAOYSA-N | |
yttrium | |
88.906 | |
23993 | |
Foil, 0.25mm (0.01 in.) thick |
50 x 50mm | |
7440-65-5 | |
4.47g/cm3 | |
Y | |
yttrium, elemental, yttrium-89, ion, ytrio, hydride, unii-58784xqc3y, yttrium, metal and compounds, ingot, foil, element | |
[Y] | |
Odorless | |
Mylar under Argon | |
CHEBI:33331 | |
Gray |
Chemical Identifiers
7440-65-5 | |
88.906 | |
VWQVUPCCIRVNHF-UHFFFAOYSA-N | |
23993 | |
yttrium |
Y | |
MFCD00011468 | |
yttrium, elemental, yttrium-89, ion, ytrio, hydride, unii-58784xqc3y, yttrium, metal and compounds, ingot, foil, element | |
CHEBI:33331 | |
[Y] |