missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Yttrium foil, 0.1mm (0.004 in.) thick, 99.9% (REO)

Manufacturer:  Alfa Aesar™ 12442FI

 View more versions of this product

Catalog No. AA12442FI



Description

Description

  • Packaged under argon
Specifications

Specifications

Yttrium foil
50 x 50mm
7440-65-5
Y
yttrium, elemental, yttrium-89, ion, ytrio, hydride, unii-58784xqc3y, yttrium, metal and compounds, ingot, foil, element
[Y]
Odorless
Mylar under Argon
CHEBI:33331
99.9%
≈0.28g/25x25mm
0.1mm (0.004 in.) thick
(REO)
MFCD00011468
VWQVUPCCIRVNHF-UHFFFAOYSA-N
yttrium
88.906
23993
Foil

Chemical Identifiers

7440-65-5
88.906
VWQVUPCCIRVNHF-UHFFFAOYSA-N
23993
yttrium
Y
MFCD00011468
yttrium, elemental, yttrium-89, ion, ytrio, hydride, unii-58784xqc3y, yttrium, metal and compounds, ingot, foil, element
CHEBI:33331
[Y]
Safety and Handling

Safety and Handling

TSCA : TSCA

storageNote1 :

  • Packaged under argon

Documents