Please login to your online account to display your discounted pricing

Alfa Aesar™ Yttrium aluminum oxide, 99.99% (metals basis)

Can be used as a sputtering target to prepare bubble memory devices

Chemical Identifiers

Chemical Name or Material Yttrium aluminum oxide
CAS 12005-21-9
Formula Weight 593.62
MDL Number MFCD00213958
Synonym Yttrium aluminum garnet; YAG
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3624114

  SDS
alfa aesar™
3624114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3624122

  SDS
alfa aesar™
3624122
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 593.62
Chemical Name or Material Yttrium aluminum oxide
TSCA TSCA
Particle Size 3 to 12mm
Physical Form Fused Lump
Refractive Index 1.833
Assay 99.99%
Assay Percent Notes (metals basis)
MDL Number MFCD00213958
CAS 12005-21-9
Synonym Yttrium aluminum garnet; YAG
Melting Point 1760°C
Quantity 25g
Density 4.65g/mL