Please login to your online account to display your discounted pricing

Yttrium acetate hydrate 99.9%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC392211000

 View more versions of this product

Catalog No. AC392211000

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder and Lumps
Formula Weight 266.04
Quantity 100g
Color White
Purity Grade For trace metal basis analysis
Assay Percent Range 99.90%
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD00150128
Infrared Spectrum Authentic
Total Impurity 0.1% max.
Packaging Glass bottle
CAS 207801-28-3
Molecular Formula C6H9O6Y·xH2O
Chemical Name or Material Yttrium acetate hydrate
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Yttrium acetate hydrate
CAS 207801-28-3
Purity Grade For trace metal basis analysis
Assay Percent Range 99.90%
Molecular Formula C6H9O6Y·xH2O
Formula Weight 266.04
MDL Number MFCD00150128
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents