missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Yttrium, AAS standard solution, Y 1000μg/mL, Alfa Aesar™, Specpure™

Manufacturer:  Alfa Aesar 88117AE

 View more versions of this product

Catalog No. AA88117AE



Specifications

Specifications

Yttrium
Y 1000;mug/mL
MFCD00011468
[N+](=O)(O)[O-]
63.012
CHEBI:48107
Characteristic
7697-37-2
HNO3
GRYLNZFGIOXLOG-UHFFFAOYSA-N
nitric acid
944
Liquid
100mL

Chemical Identifiers

7697-37-2
63.012
GRYLNZFGIOXLOG-UHFFFAOYSA-N
CHEBI:48107
[N+](=O)(O)[O-]
HNO3
MFCD00011468
944
nitric acid
Safety and Handling

Safety and Handling

H314-H318: Causes severe skin burns and eye damage. Causes serious eye damage.

DOTInformation : DOT Class: 8, Packaging Group: III

EINECSNumber : 231-714-2

SDS Documents