missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ytterbium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Manufacturer:  Spex Certiprep Llc PLYB22Y

 View more versions of this product

Catalog No. PLYB2-2Y


 

Disclaimers:

Product intended for laboratory use only


Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Specifications

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Yb1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Description & Specifications

Specifications

Ytterbium Standard
0.1,2.0,97.9
Assurance Grade
Triple-leached LDPE bottle
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
125mL
7440-64-4,7697-37-2,7732-18-5
0.1,2.0,97.9
Matrix    Concentration    Cat. No.
2-5% HNO3    1000μg/mL    PLYB2-2Y, -2X    10,000    PLYB2-3Y, -3X
Assurance Certificate of Analysis reports up to 39 elements at ppb levels.
2% HNO3
AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

Chemical Identifiers

AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Ytterbium Standard

Safety and Handling

DOTInformation : Hazardous

Recommended Storage : Ambient