missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ytterbium sputtering target, 50.8mm dia x 3.18mm thick

$1,810.19 - $1,810.19

Chemical Identifiers

CAS 7440-64-4
MDL Number MFCD00011286
Chemical Name or Material Ytterbium sputtering target, 50.8mm dia. x 3.18mm thick
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA45736KS SDS Alfa Aesar™
45736KS
1 ea. Each for $1,810.19
Specifications Description & Specifications

Chemical Identifiers

7440-64-4
MFCD00011286
Ytterbium sputtering target, 50.8mm dia. x 3.18mm thick

Specifications

1 ea.
Odorless
Ytterbium sputtering target, 50.8mm dia. x 3.18mm thick
7440-64-4
MFCD00011286

Safety and Handling

TSCA : TSCA