missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ytterbium sputtering target, 50.8mm dia x 1.59mm thick

$1,614.70 - $1,614.70

Chemical Identifiers

CAS 7440-64-4
MDL Number MFCD00011286
Chemical Name or Material Ytterbium sputtering target, 50.8mm dia. x 1.59mm thick
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA45620KS SDS Alfa Aesar™
45620KS
1 ea. Each for $1,614.70
Specifications Description & Specifications

Chemical Identifiers

7440-64-4
MFCD00011286
Ytterbium sputtering target, 50.8mm dia. x 1.59mm thick

Specifications

1 ea.
Odorless
Ytterbium sputtering target, 50.8mm dia. x 1.59mm thick
7440-64-4
MFCD00011286

Safety and Handling

TSCA : TSCA