Learn More
Ytterbium pieces, sublimed, 99.9% (REO), Thermo Scientific Chemicals
May contain up to 0.15% Calcium
Supplier: Thermo Scientific Chemicals 04343106
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Ytterbium | |
7440-64-4 | |
5 g | |
Yb | |
unii-mnq4o4wsi1, hydride, mnq4o4wsi1, foil, 3n, yterbio, ingot, powder, ii ion, chips, 3n, ingot, 3n | |
[Yb] | |
173.05 | |
CHEBI:33381 | |
Odorless |
May contain up to 0.15% Calcium | |
Silver to Gray | |
99.9% | |
MFCD00011286 | |
NAWDYIZEMPQZHO-UHFFFAOYSA-N | |
ytterbium | |
23992 | |
173.04 | |
Silver Gray pieces |
Chemical Identifiers
7440-64-4 | |
173.05 | |
NAWDYIZEMPQZHO-UHFFFAOYSA-N | |
23992 | |
ytterbium |
Yb | |
MFCD00011286 | |
unii-mnq4o4wsi1, hydride, mnq4o4wsi1, foil, 3n, yterbio, ingot, powder, ii ion, chips, 3n, ingot, 3n | |
CHEBI:33381 | |
[Yb] |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : DOT Class: 4.3, Packaging Group: II
EINECSNumber : 231-173-2
RTECSNumber : ZG1925000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only