missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ytterbium pieces, sublimed, 99.9% (REO), Alfa Aesar™

May contain up to 0.15% Calcium

Manufacturer:  Alfa Aesar 4343122

 View more versions of this product

Catalog No. AA4343122



Specifications

Specifications

Ytterbium
7440-64-4
Yb
Silver Gray pieces
NAWDYIZEMPQZHO-UHFFFAOYSA-N
ytterbium
23992
173.04
Odorless
100g
May contain up to 0.15% Calcium
99.9%
MFCD00011286
chips, 3n
[Yb]
173.054
CHEBI:33381
Sublimed Pieces
Silver to Gray

Chemical Identifiers

7440-64-4
173.054
NAWDYIZEMPQZHO-UHFFFAOYSA-N
23992
ytterbium
Yb
MFCD00011286
chips, 3n
CHEBI:33381
[Yb]
Safety and Handling

Safety and Handling

H261: In contact with water releases flammable gas.

DOTInformation : DOT Class: 4.3, Packaging Group: II

EINECSNumber : 231-173-2

Documents