Please login to your online account to display your discounted pricing

Alfa Aesar™ Ytterbium(III) trifluoromethanesulfonate hydrate, 98%

Chemical Identifiers

Chemical Name or Material Ytterbium(III) trifluoromethanesulfonate hydrate
Formula Weight 620.24 (Anhydrous)
MDL Number MFCD03703499
Synonym Ytterbium(III) triflate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4031406

  SDS
alfa aesar™
4031406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4031414

  SDS
alfa aesar™
4031414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 620.24 (Anhydrous)
Chemical Name or Material Ytterbium(III) trifluoromethanesulfonate hydrate
Physical Form Powder
Assay 98%
MDL Number MFCD03703499
Synonym Ytterbium(III) triflate
Quantity 5g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.