missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ytterbium(III) sulfate octahydrate, REacton™, 99.9% (REO)

Manufacturer:  Alfa Aesar™ 1457218

 View more versions of this product

Catalog No. AA1457218



Specifications

Specifications

Ytterbium(III) sulfate octahydrate
Reacton™
(REO)
MFCD00149887
MDOSIOHMOWGVIM-UHFFFAOYSA-H
ytterbium(3+);trisulfate;octahydrate
25022290
Crystalline
10034-98-7
99.9%
H16O20S3Yb2
14,10105
O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Yb+3].[Yb+3]
778.396
778.38 (634.26 Anhydrous)
50g

Chemical Identifiers

10034-98-7
778.396
MDOSIOHMOWGVIM-UHFFFAOYSA-H
ytterbium(3+);trisulfate;octahydrate
H16O20S3Yb2
MFCD00149887
25022290
O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Yb+3].[Yb+3]
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents