Please login to your online account to display your discounted pricing

Ytterbium(III) oxide, 99.998% (REO), Alfa Aesar™, Reacton™

$75.95 - $280.26

Chemical Identifiers

Chemical Name or Material Ytterbium(III) oxide
CAS 1314-37-0
Physical Form Powder
Assay Percent Range 99.998% (REO)
Molecular Formula Yb2O3
Formula Weight 394.08
MDL Number MFCD00011290
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1091906

  SDS
alfa aesar
1091906
5g Each for $75.95

AA1091914

  SDS
alfa aesar
1091914
25g Each for $280.26
Description & Specifications

Specifications

Chemical Name or Material Ytterbium(III) oxide
CAS 1314-37-0
EINECS Number 215-234-0
Physical Form Powder
Assay Percent Range 99.998% (REO)
Molecular Formula Yb2O3
Formula Weight 394.08
MDL Number MFCD00011290
Quantity 5g