Please login to your online account to display your discounted pricing

Ytterbium(III) oxide, 99.99%, (trace metal basis), ACROS Organics™

$190.13 - $190.13

Chemical Identifiers

Chemical Name or Material Ytterbium(III) oxide
Name Note 99%
CAS 1314-37-0
Purity Grade For trace metal basis analysis
Assay 86.5 to 89.1% (Yb)
Assay Percent Range 99.99%
Molecular Formula O3Yb2
Linear Formula Yb2O3
Formula Weight 394.08
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC264300100

  SDS
thermo fisher scientific
AC264300100
Glass bottle 10g Each for $190.13
Description & Specifications

Specifications

Physical Form Powder
Formula Weight 394.08
Quantity 10g
Color White
Assay 86.5 to 89.1% (Yb)
EINECS Number 215-234-0
TSCA TSCA
Purity Grade For trace metal basis analysis
Density 9.17
Name Note 99%
Chemical Name or Material Ytterbium(III) oxide
Packaging Glass bottle
Assay Percent Range 99.99%
Molecular Formula O3Yb2
Linear Formula Yb2O3
CAS 1314-37-0
Total Trace Metal Impurities 100ppm max.
Health Hazard 1 HYGROSCOPIC

for ultra trace analysis

Safety and Handling

HYGROSCOPIC