Please login to your online account to display your discounted pricing

Ytterbium(III) oxide, 99.9% (REO), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Ytterbium oxide
CAS 1314-37-0
Physical Form 3 to 12mm Sintered Lump
Assay Percent Range 99.9% (REO)
Molecular Formula Yb2O3
Formula Weight 394.08
MDL Number MFCD00011290
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3625214

  SDS
alfa aesar
3625214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3625222

  SDS
alfa aesar
3625222
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 394.08
Physical Form 3 to 12mm Sintered Lump
Quantity 25g
Density 9.2g/mL
EINECS Number 215-234-0
Molecular Formula Yb2O3
CAS 1314-37-0
MDL Number MFCD00011290
Chemical Name or Material Ytterbium oxide
Assay Percent Range 99.9% (REO)