missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ytterbium(III) nitrate hydrate, REacton™, 99.99% (REO)

Chemical Identifiers

Formula Weight 359.05 (Anhydrous)
MDL Number MFCD00149883
UN Number UN1477
Merck Index 14,10105
Chemical Name or Material Ytterbium(III) nitrate hydrate
Grade Reacton™
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1119609 SDS Alfa Aesar™
1119609
10g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA1119618 SDS Alfa Aesar™
1119618
50g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

359.05 (Anhydrous)
UN1477
Ytterbium(III) nitrate hydrate
MFCD00149883
14,10105
Reacton™

Specifications

Crystalline Aggregates
MFCD00149883
14,10105
White
Ytterbium(III) nitrate hydrate
Reacton™
359.05 (Anhydrous)
UN1477
Hygroscopic
10g
99.99%, (REO)

Safety and Handling

GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P221-P210-P220-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Keep/Store away from clothing/combustible materials.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.

RTECSNumber : ZG2800000

TSCA : TSCA