missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ytterbium(III) nitrate hydrate, 99.9% (REO)

Manufacturer:  Alfa Aesar™ 1290109

 View more versions of this product

Catalog No. AA1290109



Specifications

Specifications

Ytterbium(III) nitrate hydrate
MFCD00149883
14,10105
Crystalline
Hygroscopic
10g
(REO)
UN1477
359.05 (Anhydrous)
99.9%
White

Chemical Identifiers

MFCD00149883
Safety and Handling

Safety and Handling

GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P221-P210-P220-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Keep/Store away from clothing/combustible materials.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.

RTECSNumber : ZG2800000

TSCA : TSCA

Documents