missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ytterbium(III) chloride, ultra dry, 99.99% (REO), Alfa Aesar™

Chemical Identifiers

CAS 10361-91-8
Physical Form Beads
Assay Percent Range 99.99% (REO)
Molecular Formula YbCl3
Formula Weight 279.4
MDL Number MFCD00011288
Chemical Name or Material ultra dry
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4065306 SDS Alfa Aesar
4065306
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA4065314 SDS Alfa Aesar
4065314
25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

10361-91-8
99.99% (REO)
279.4
ultra dry
Beads
YbCl3
MFCD00011288

Specifications

279.4
Off-white
99.99% (REO)
10361-91-8
MFCD00011288
5g
Beads
Odorless
YbCl3
ultra dry