Please login to your online account to display your discounted pricing

Alfa Aesar™ Ytterbium(III) chloride hydrate, REacton™, 99.9% (REO)

$54.19 - $197.03

Chemical Identifiers

Chemical Name or Material Ytterbium(III) chloride hydrate
CAS 19423-87-1
Purity Grade Reacton™
Formula Weight 279.40 (Anhydrous)
MDL Number MFCD00149885
Merck Index 14,10105
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1119309

 
alfa aesar™
1119309
10g Each for $54.19

AA1119318

 
alfa aesar™
1119318
50g Each for $197.03
Description & Specifications

Specifications

Formula Weight 279.40 (Anhydrous)
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Ytterbium(III) chloride hydrate
TSCA TSCA
Odor Odorless
Purity Grade Reacton™
Physical Form Wet Crystalline Aggregates
Assay 99.9%
Sensitivity Hygroscopic
Assay Percent Notes (REO)
MDL Number MFCD00149885
CAS 19423-87-1
Merck Index 14,10105
Color Green
Melting Point 150° to 155°C
Quantity 10g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Density 2.58g/mL
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.