Please login to your online account to display your discounted pricing

Ytterbium(III) chloride hydrate, 99.99% (REO), REacton™, Alfa Aesar™

$394.77 - $394.77

Chemical Identifiers

CAS 19423-87-1
Physical Form Crystalline Aggregates
Assay Percent Range 99.99% (REO)
Molecular Formula YbCl3·xH2O (x≈6)
Formula Weight 279.4
MDL Number MFCD00149885
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1119409

  SDS
alfa aesar
1119409
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1119418

  SDS
alfa aesar
1119418
50g Each for $394.77
Description & Specifications

Specifications

Formula Weight 279.4
Quantity 10g
Color White
Physical Form Crystalline Aggregates
CAS 19423-87-1
Odor Odorless
Molecular Formula YbCl3·xH2O (x≈6)
MDL Number MFCD00149885
Assay Percent Range 99.99% (REO)