missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ytterbium(III) chloride hexahydrate, 99.9%, (trace metal basis), Thermo Scientific™
Supplier: Thermo Scientific Chemicals 194430010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
for ultra trace analysisSpecifications
Ytterbium(III) chloride hexahydrate | |
1g | |
Cl3Yb·6H2O | |
MFCD00149885 | |
14, 10105 | |
387.48 | |
99.9% | |
Glass bottle |
10035-01-5 | |
99.90% | |
YbCl3·6H2O | |
16,384 | |
Solubility in water: soluble. Other solubilities: soluble in absolute alcohol | |
387.48 | |
Trace Metal Basis |
Chemical Identifiers
10035-01-5 | |
Cl3Yb·6H2O | |
387.48 | |
MFCD00149885 |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
GHS Signal Word: Warning
RUO â Research Use Only