missing translation for 'onlineSavingsMsg'
Learn More

Ytterbium(III) chloride, 35% min w/w aq. soln., 99.9% (REO)

CAS: 10361-91-8 | YbCl3

Supplier:  Thermo Scientific Chemicals 042816AK

 View more versions of this product

Catalog No. AA42816AK

Only null left
Explore available promotions

Description

Description

Applied with NiCl2 to form a very effective catalyst for the reductive dehalogenation of aryl halides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ytterbium (III) chloride
Liquid
(REO)
MFCD00011288
279.40 (Anhydrous)
35% min w/w aq. soln.
1.48 g/mL at 30°C
10361-91-8
250 mL
YbCl3
Soluble in water.
99.9%
Odorless

Chemical Identifiers

10361-91-8
YbCl3
MFCD00011288
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 233-800-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only