missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ytterbium(III) bromide hydrate, REacton™, 99.99% (REO)

Manufacturer:  Alfa Aesar™ 4405709

 View more versions of this product

Catalog No. AA4405709



Specifications

Specifications

Ytterbium (III) bromide hydrate
(REO)
MFCD00049613
412.77 (Anhydrous)
99.99%
10g
226944-34-9
YbBr3·xH2O
acmc-20aj9h
Lumps
Reacton™

Chemical Identifiers

226944-34-9
YbBr3·xH2O
MFCD00049613
acmc-20aj9h
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 237-350-0

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents