Please login to your online account to display your discounted pricing

Alfa Aesar™ Ytterbium(III) acetate hydrate, REacton™, 99.9% (REO)

Chemical Identifiers

Chemical Name or Material Ytterbium(III) acetate hydrate
CAS 15280-58-7
Purity Grade Reacton™
Formula Weight 350.16 (Anhydrous)
MDL Number MFCD00150127
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1457618

  SDS
alfa aesar™
1457618
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 350.16 (Anhydrous)
Assay 99.9%
Quantity 50g
Assay Percent Notes (REO)
Purity Grade Reacton™
CAS 15280-58-7
Odor Odorless
MDL Number MFCD00150127
Physical Form Crystalline Aggregates
Chemical Name or Material Ytterbium(III) acetate hydrate
TSCA TSCA