missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ytterbium(III) 2,4-pentanedionate, REacton™, 99.9% (REO)

Manufacturer:  Alfa Aesar™ 1315909

 View more versions of this product

Catalog No. AA1315909



Specifications

Specifications

Ytterbium(III) 2,4-pentanedionate
White
14284-98-1
C15H21O6Yb
ytterbium-2,4-pentanedionate, ytterbium acetyl acetonate
CC(=CC(=O)C)[O-].CC(=CC(=O)C)[O-].CC(=CC(=O)C)[O-].[Yb+3]
470.381
470.37
99.9%
Odorless
10g
(REO)
MFCD00013508
CVEKUGFVEWLJFN-LNTINUHCSA-K
(Z)-4-oxopent-2-en-2-olate;ytterbium(3+)
14455610
Powder
Reacton™

Chemical Identifiers

14284-98-1
470.381
CVEKUGFVEWLJFN-LNTINUHCSA-K
14455610
CC(=CC(=O)C)[O-].CC(=CC(=O)C)[O-].CC(=CC(=O)C)[O-].[Yb+3]
C15H21O6Yb
MFCD00013508
ytterbium-2,4-pentanedionate, ytterbium acetyl acetonate
(Z)-4-oxopent-2-en-2-olate;ytterbium(3+)
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents