missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ytterbium foil, 1.0mm (0.04 in.) thick, 99.9% (REO)

Manufacturer:  Alfa Aesar™ 10426FI

 View more versions of this product

Catalog No. AA10426FI



Description

Description

  • Packaged under argon
Specifications

Specifications

Ytterbium foil
50 x 50 mm
7440-64-4
Yb
chips, 3n, foil, 3n, hydride, ii ion, ingot, ingot, 3n, mnq4o4wsi1, powder, unii-mnq4o4wsi1, yterbio
[Yb]
Odorless
Mylar under Argon
CHEBI:33381
99.9%
≈4.4g/25x25mm
1.0mm (0.04 in.) thick
(REO)
MFCD00011286
NAWDYIZEMPQZHO-UHFFFAOYSA-N
ytterbium
173.054
23992
Foil

Chemical Identifiers

7440-64-4
173.054
NAWDYIZEMPQZHO-UHFFFAOYSA-N
23992
ytterbium
Yb
MFCD00011286
chips, 3n, foil, 3n, hydride, ii ion, ingot, ingot, 3n, mnq4o4wsi1, powder, unii-mnq4o4wsi1, yterbio
CHEBI:33381
[Yb]
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

EINECSNumber : 231-173-2

TSCA : Yes

storageNote1 :

  • Packaged under argon

Recommended Storage : Ambient temperatures

Documents