Please login to your online account to display your discounted pricing

Ytterbium, 99.9%, (trace metal basis), -12mm, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  297510050

 View more versions of this product

Catalog No. AC297510050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Purity Grade Trace Metal Basis
Color Gray with metal luster
Physical Form Pieces With Metal Luster
Formula Weight 173.04
Quantity 5g
Merck Index 15, 10303
TSCA TSCA
RTECS Number ZG1925000
EINECS Number 231-173-2
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula Yb
Chemical Name or Material Ytterbium
CAS 7440-64-4
Linear Formula Yb
Health Hazard 1 MOISTURE SENSITIVE
MDL Number MFCD00011286
Total Trace Metal Impurities 0.1% max.
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Name Note 99.9%, -12 mm
Assay Percent Range 99.9%

Specifications

Chemical Name or Material Ytterbium
Name Note 99.9%, -12 mm
CAS 7440-64-4
Purity Grade Trace Metal Basis
Assay Percent Range 99.9%
Molecular Formula Yb
Linear Formula Yb
Formula Weight 173.04
MDL Number MFCD00011286
Accessories

MOISTURE SENSITIVE

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.

SDS & Safety Documents