Please login to your online account to display your discounted pricing

YSI™ Zobell ORP Calibration Solution

For calibrating ORP/Redox on several YSI instruments

$61.19 - $61.19

Specifications

Applications Calibrating ORP/Redox on YSI™ instruments
Filling Solution Type ORP calibration solution
Includes Temperature chart with corresponding ORP values is on the side of the container
Type Calibration Solution
Volume (Metric) Sample 119mL
View More Specs
Products
Catalog Number Mfr. No. Price Quantity    

15-177-25

 
ysi™
YSI061320*
Each for $61.19
Description & Specifications

Specifications

Applications Calibrating ORP/Redox on YSI™ instruments
Filling Solution Type ORP calibration solution
Includes Temperature chart with corresponding ORP values is on the side of the container
Type Calibration Solution
Volume (Metric) Sample 119mL
Warranty 2 Years

  • A convenient temperature chart with corresponding ORP values is on the side of the container for simple calibration value inputs
  • Concentrated power makes 125mL of calibration standard
  • Plastic bottle (125mL)

Expiration Dates:
Unmixed: 2 years
Mixed: 6 months