YSI™ Probe Service Kits for YSI Dissolved Oxygen Probes
For replenishing oxygen sensor on YSI Dissolved Oxygen Probes
Supplier: YSI™ 5794
Description
- Membrane Kits include 150 standard, 1mil PTFE membranes or 150 high-sensitivity, 0.5mil PTFE membranes, in 10 packs of 50
- Membrane/KCI Kits include 30 standard or high-sensitivity membranes, 1 oz. (30mL) dropper bottle with KCI crystals for adding distilled water and a probe contour tool
- For use with probes 13-299-43 (Model 5739), 13-298-80 (Model 5750), 15-176-50 (Model 5718), 13-298-16 (Model 5300 Biological Oxygen Monitor), and 13-298-56, -57, or -62 (Model 55 Handheld Dissolved Oxygen Meters)
Specifications
YSI™ 55, 5739, 5750, 5718, 6030 and 6562 DO Probe | |
10/pk. | |
10/pk. |
High Sensitivity Membrane Kit: 150 membranes | |
DO |