YSI™ Probe Service Kits for YSI Dissolved Oxygen Probes
For replenishing oxygen sensor on YSI Dissolved Oxygen Probes
Manufacturer:  YSI™ YSI5775
 View more versions of this product
Catalog No. 13-299-16
Specifications Description & Specifications
Specifications
5718, 5739, 5750, 5300 and Rapid Pulse Probes
DO
Bottle of electrolyte, 2 O-rings, 30 1mL FEP Fluoropolymer Film Membranes
Membrane Kit