missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
YSI™ pHotoFlex™ Colorimeters
Waterproof, multiparameter colorimeters are ideal for laboratory and field testing
$1,826.06 - $3,844.42
Specifications
Certifications/Compliance | CE, ETLus, cETL |
---|---|
Dimensions (L x W x H) | 9.3 x 3.4 x 4.6 in. (23.6 x 8.6 x 11.7cm) |
IP Rating | IP67 |
Warranty | 2 Year |
Weight (English) | 1.3 lb. |
Includes: pHotoFlex pH/SET or STD/SET: pHotoFlex pH or STD colorimeter, field case with field tray, vials, pH electrode (with pH/SET), 2 each 10 mL and 2 mL syringe, empty cuvettes 16+28 mm, cleaning tissues, stand, 2 empty buffer bottles, beaker, screw driver, PC-software LSdata, and RS232 PC cable
Catalog Number | Mfr. No. | Data Storage | Includes | Product Type | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Data Storage | Includes | Product Type | Price | Quantity | |||
15-177-700 |
YSI™
YSI251105Y |
100 Data sets | Colorimeter, Batteries | pHotoFlex STD colorimeter only |
Each for $1,826.06
|
|
15-177-701 |
YSI™
YSI251100Y |
1000 Data sets | Colorimeter, Batteries | pHotoFlex pH colorimeter only |
Each for $2,488.42
|
|
15-177-703 |
YSI™
YSI251205Y |
100 Data sets | LED filter colorimeter, field case with field tray, vials, pH electrode (with pH/SET), 2 each 10 mL and 2 mL syringe, empty cuvettes 16+28 mm, cleaning tissues, stand, 2 empty buffer bottles, beaker, screw driver, PC-software LSdata, and RS232 PC cable | pHotoFlex STD colorimeter set |
Each for $3,220.66
|
|
15-177-702 |
YSI™
YSI251200Y |
1000 Data sets | LED filter, case, tray, stands, vials, pH electrode, syringes, cuvettes, cleaning tissues, buffer bottles, beaker, screwdriver, Lsdata software, RS232 PC cable, BNC connector | pHotoFlex pH colorimeter set |
Each for $3,844.42
|
|
Description
- Large, backlit display
- Data logging enables exporting data to computer using LSdata software
- User-defined programs
- Flexible, fixed adapter with flip design
pHotoFlex STD
- For colorimeter only
- Data storage; 100 data sets
- No. of tests: 85
- For colorimeter and pH electrode
- Data storage; 1,000 data sets
- No. of tests: ∽138 tests
- Offers BNC connection
Order Info
- Dimensions : 17H x 86W x 236mm L (4.6 x 3.4 x 9.3 in.)
- Weight : 0.6kg (1.3 lb.)
Certifications
CE Marked, ETLus, cETL, IP67 waterproof
Warranty
2-year
Specifications
CE, ETLus, cETL | |
IP67 | |
1.3 lb. | |
Backlit LCD |
9.3 x 3.4 x 4.6 in. (23.6 x 8.6 x 11.7cm) | |
2 Year | |
0.6kg | |
0° to 50°C |