missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

YSI™ pHotoFlex™ Colorimeters

Waterproof, multiparameter colorimeters are ideal for laboratory and field testing

Manufacturer:  YSI™ YSI251200Y

 View more versions of this product

Catalog No. 15-177-702



Includes: pHotoFlex pH/SET or STD/SET: pHotoFlex pH or STD colorimeter, field case with field tray, vials, pH electrode (with pH/SET), 2 each 10 mL and 2 mL syringe, empty cuvettes 16+28 mm, cleaning tissues, stand, 2 empty buffer bottles, beaker, screw driver, PC-software LSdata, and RS232 PC cable

Description

Description

  • Large, backlit display
  • Data logging enables exporting data to computer using LSdata software
  • User-defined programs
  • Flexible, fixed adapter with flip design

pHotoFlex STD

  • For colorimeter only
  • Data storage; 100 data sets
  • No. of tests: 85
pHotoFlex pH
  • For colorimeter and pH electrode
  • Data storage; 1,000 data sets
  • No. of tests: ∽138 tests
  • Offers BNC connection

Order Info

  • Dimensions : 17H x 86W x 236mm L (4.6 x 3.4 x 9.3 in.)
  • Weight : 0.6kg (1.3 lb.)

Certifications

CE Marked, ETLus, cETL, IP67 waterproof

Warranty

2-year

Specifications

Specifications

9.3 x 3.4 x 4.6 in. (23.6 x 8.6 x 11.7cm)
Backlit LCD
11.7cm
23.6cm
0.6kg
8.6cm
2 Year
0° to 50°C
1000 Data sets
pHotoFlex pH colorimeter set
4.6 in.
9.3 in.
1.3 lb.
3.4 in.
IP67
LED filter, case, tray, stands, vials, pH electrode, syringes, cuvettes, cleaning tissues, buffer bottles, beaker, screwdriver, Lsdata software, RS232 PC cable, BNC connector
CE, ETLus, cETL
Documents