missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

YSI™ pH Probes for Pro Series Portable Meters

Choose from lab-grade and field models.

Manufacturer:  YSI™ YSI605101

 View more versions of this product

Catalog No. 15-177-460



Description

Description

  • For use with Pro Series Portable Meters
  • Without cable
  • Requires cable/probe assembly
Specifications

Specifications

Pro series meter
pH
Without cable
pH probe
Documents