missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

YSI™ pH Probes for Pro Series Portable Meters

Choose from lab-grade and field models.

Manufacturer:  YSI™ YSI605107

 View more versions of this product

Catalog No. 15-177-500



Includes: 1m cable and connector

Description

Description

  • For use with Pro Series Portable Meters
  • With 1m cable and connector
Specifications

Specifications

Single
Pro series meter
pH
pH/ATC lab-grade probe
1m
39.3 in.
Cable and connector
Documents